Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZING™ Enterprises Eco Safety Projecting Sign, Warning BioHazard
ZING Enterprises Eco Safety Projecting sign is available as a V style or L style from recycled plastic.
Supplier: ZING™ Enterprises 2609
Description
Features:
- Clearly communicates critical safety information
- Two-sided projecting design significantly increases visibility, and is ideal for designating important workplace locations
- Made in the USA from 70% post-consumer recycled plastic
- Two mounting holes and adhesive tabs are included for easy installation
- Recycled materials support sustainability initiatives and LEED credits
Specifications
Plastic | |
Corner Holes | |
7 in. | |
English | |
Warning Biohazard |
1 Ea. | |
Communication | |
2.5 in. | |
7 x 2.5 in. | |
White |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction