Learn More
ZING™ Enterprises Eco Safety Sign NOTICE Protective Clothing Required In This Area
The sign is available in 2 sizes, 7 in. x 10 in., and 10 in. x 14 in., and three eco-friendly formats.
Manufacturer:  ZING™ Enterprises 20061S
 View more versions of this product
Catalog No. 18-002-511
Add to cart
Description
Description
  • Plastic signs are made from a minimum of 70% post-consumer recycled materials and are printed with UV-resistant inks. The signs are pre-drilled for easy mounting, and include adhesive mounting tabs. Impact-resistant plastic is suitable for indoor or outdoor use.
  • Aluminum signs are made from 94% recycled materials. The signs are printed with UV-resistant inks, and are suitable for indoor and outdoor use. The signs are pre-drilled for easy mounting
  • Adhesive signs are made from a minimum of 70% post-industrial polystyrene, and are resistant to solvents, moisture, scuffing, tears and UV exposure
  • All Zing signs are made in the USA from recycled materials. Recycled materials support sustainability initiatives and LEED credits. The sign meets OSHA and ANSI standards. Signs are sold 1/pack.
Specifications
Specifications
Black, Blue, White
Self-Adhesive
10 in. x 14 in.
Self-Adhesive
10 in.
14 in.
English
1
14 in.
Communication
NOTICE Protective Clothing Required In This Area