Learn More
ZING™ Enterprises Eco Safety Tag Center
Features 8 durable custom hooks that accommodate up to 10 tags per hook.
Manufacturer:  ZING™ Enterprises 2670
Catalog No. 18-002-855
Add to cart
Description
Description
ZING Eco Safety Tag Center is designed to organize and store a variety of tags commonly used in safety applications such as lockout tagout or equipment inspections. Tag center features 8 durable custom hooks that accommodate up to 10 tags per hook. Station is made from heavy-duty impact resistant recycled PETG that is high visible green to attract attention and encourage use. Recycled materials support sustainability initiatives and LEED credits. Station includes pre-drilled holes for easy mounting (hardware not included) and measures 15 in.H x 15 in.W. Made in the USA. (ZING Safety Tags Sold Separately).
Specifications
Specifications
Green
Plastic
15.5 in. x 15.5 in.
Corner Holes
15.5 in.
15.5 in.
English
1
15.5 in.
Safety
Lockout