Learn More
ZING™ Enterprises Eco Single Box Glove Dispenser
Available in clear or white, and in single, double, or triple box sizes.
Manufacturer:  ZING™ Enterprises 7220
 View more versions of this product
Catalog No. 18-002-987
Add to cart
Description
Description
Eco Single Box Glove Dispenser conveniently stores and organizes glove boxes on a wall, cabinet or other surface in either the vertical or horizontal position. Glove dispenser is made from high quality .150 in. thick 100% recycled PETG providing superior durability and crack resistance over other materials like acrylic. Dispenser is manufactured using high content recycled materials that support sustainability initiatives and LEED Credits. Single box dispenser with clear recycled plastic measures 10.5 in.L x 6 in.W x 4 in.H. Made in the USA. (Optional magnet mount, ZING 7226, can be purchased separately).
Magnet mounts can be used with Eco Gove Dispensers to allow easy placement of glove dispensers on metal surfaces including filing cabinets, storage cabinets, metal doors, and other metal surfaces. One magnet required for single, two for double and three for triple dispensers. Made in the USA.
Specifications
Specifications
Clear
10.5 in. x 6 in.
Corner Holes
10.5 in.
Plastic
Glove Dispenser
Dispenser
1
6 in.