Learn More
ZING™ Enterprises Eco Single Box Glove Dispenser
Available in clear or white, and in single, double, or triple box sizes.
$10.30 - $84.44
Specifications
For Use With (Application) Dispenser
No. per Pack 1
View More Specs
Products 7
Catalog Number Mfr. No. Color Dimensions (L x W) Material Mounting Price Quantity & Availability  
Catalog Number Mfr. No. Color Dimensions (L x W) Material Mounting Price Quantity & Availability  
18-002-987
View Documents
ZING™ Enterprises
7220
Clear 10.5 in. x 6 in. Plastic Corner Holes Each for $50.47
Add to cart
18-002-988
View Documents
ZING™ Enterprises
7221
Clear 12 in. x 10.5 in. Plastic Corner Holes Each for $64.89
Add to cart
18-002-989
View Documents
ZING™ Enterprises
7222
Clear 16 in. x 10.5 in. Plastic Corner Holes Each for $81.37
Add to cart
18-002-990
View Documents
ZING™ Enterprises
7223
White 10.5 in. x 6 in. Plastic Corner Holes Each for $51.00
Add to cart
18-002-991
View Documents
ZING™ Enterprises
7224
White 12 in. x 10.5 in. Plastic Corner Holes Each for $66.00
Add to cart
18-002-992
View Documents
ZING™ Enterprises
7225
White 16 in. x 10.5 in. Plastic Corner Holes Each for $84.44
Add to cart
18-002-993
View Documents
ZING™ Enterprises
7226
Black 10 in. x 5 in. Magnet Magnetic Each for $10.30
Add to cart
Description
Description
Eco Single Box Glove Dispenser conveniently stores and organizes glove boxes on a wall, cabinet or other surface in either the vertical or horizontal position. Glove dispenser is made from high quality .150 in. thick 100% recycled PETG providing superior durability and crack resistance over other materials like acrylic. Dispenser is manufactured using high content recycled materials that support sustainability initiatives and LEED Credits. Single box dispenser with clear recycled plastic measures 10.5 in.L x 6 in.W x 4 in.H. Made in the USA. (Optional magnet mount, ZING 7226, can be purchased separately).
Magnet mounts can be used with Eco Gove Dispensers to allow easy placement of glove dispensers on metal surfaces including filing cabinets, storage cabinets, metal doors, and other metal surfaces. One magnet required for single, two for double and three for triple dispensers. Made in the USA.
Specifications
Specifications
Dispenser
1