Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Econazole nitrate, Alfa Aesar™

Click to view available options
Quantity:
5g
25g
24169-02-6
C18H16Cl3N3O4
444.693
MFCD00058160
DDXORDQKGIZAME-UHFFFAOYSA-N
econazole nitrate,ifenec,epi-pevaryl,gyno-pevaryl,econazole nitrate salt,pevaryl,+--econazole nitrate,spectazole,econazolum nitrate,gyno-pevaryl 150
68589
1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
C1=CC(=CC=C1COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]
24169-02-6
C18H16Cl3N3O4
444.693
MFCD00058160
DDXORDQKGIZAME-UHFFFAOYSA-N
econazole nitrate,ifenec,epi-pevaryl,gyno-pevaryl,econazole nitrate salt,pevaryl,+--econazole nitrate,spectazole,econazolum nitrate,gyno-pevaryl 150
68589
1-[2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl]imidazole;nitric acid
C1=CC(=CC=C1COC(CN2C=CN=C2)C3=C(C=C(C=C3)Cl)Cl)Cl.[N+](=O)(O)[O-]
Melting Point 158°C to 163°C
Packaging Plastic bottle
Quantity 5g
Formula Weight 444.70
Physical Form Powder
Chemical Name or Material Econazole nitrate