Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Econazole Nitrate, BP, 99-101%, Spectrum™

Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
40 kg
Packaging:
Amber Glass Bottle
Fiber Drum
68797-31-9
C18H16Cl3N3O4
444.69
DDXORDQKGIZAME-UHFFFAOYNA-N
1-{2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl}-1H-imidazole; nitric acid
O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C=C1
68797-31-9
C18H16Cl3N3O4
444.69
DDXORDQKGIZAME-UHFFFAOYNA-N
1-{2-[(4-chlorophenyl)methoxy]-2-(2,4-dichlorophenyl)ethyl}-1H-imidazole; nitric acid
O[N+]([O-])=O.ClC1=CC=C(COC(CN2C=CN=C2)C2=CC=C(Cl)C=C2Cl)C=C1

E1107, 68797-31-9, C18H15Cl3N2O•HNO3

Spectrum™ Econazole Nitrate, BP is an antifungal medication. All Spectrum Chemical BP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Sulfated Ash 0.001
Loss on Drying 0.005
Quantity 100 g
CAS Max % 1
Percent Purity 99 to 101%