Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Iris Forceps, Sklar, 4-1/4 in., 1x2 Teeth, Half Curved, Thumb, Stainless Steel. It is sold in boxes of 12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21146
Econo Iris Forceps, Sklar, 4-1/4 in., 1x2 Teeth, Half Curved, Thumb, Stainless Steel. Econo Eye Tissue Forceps are most commonly used in ophthalmic procedures for grasping and holding tissue. This instrument is available in several different variations. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is half curved with 1x2 teeth and a length of 4-1/2 inches. It is sold in boxes of 12.
Catalog No. 50-143-7544