Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Probe with Eye, Sklar, 5 in., Blunt, Straight, Smooth, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21499
Econo Probe with Eye, Sklar, 5 in., Blunt, Straight, Smooth, Stainless Steel. Econo Probe with Eye is a slender, slightly malleable, multipurpose instrument with a bulbous tip that can be used in a variety of procedures. It may be used to explore or gauge the depth of non-visible cavities, explore lesions, manipulate tissue, or carry ligatures. Econo instruments are Sklar's floor-grade, disposables and are sold in bulk quantities. Box of 12.
Catalog No. 50-143-7658