Learn More
Sklar InstrumentsSupplier Diversity Partner Econo Graves Vaginal Speculum, Sklar, Blunt, Angled, Smooth, Medium, Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 21313
Econo Graves Vaginal Speculum, Sklar, Blunt, Angled, Smooth, Medium, Stainless Steel. Econo Graves Vaginal Speculum is a gynecological instrument used for performing pelvic examinations, Pap Smears, and examining the cervix. It is a double blade duckbill speculum inserted with the blades closed. The blades are then opened by an external lever and screw. The Graves Speculum has wider blades than the Pederson Speculum and the practitioner can best determine which type and size speculum is most appropriate for the patient. Econo instruments are Sklar's floor-grade, disposable instruments and are sold in bulk quantities. This product is a medium sized speculum and it is sold in boxes of 12.
Catalog No. 50-143-7599