Economical Allihn Condensers
Manufacturer: Tech Glass, Inc. TG2310003
 View all options for this product
Catalog No. 07-730B
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Length (English) Jacket 11.8 in.
    Includes Spherical bulbs and sealed-in water jacket
    Material Borosilicate Glass
    Length (Metric) Jacket 30cm
    Product Type Allihn Condenser
    Water jackets sealed to condensing tubes. Bulbs approximately spherical. Use No. 3 rubber stopper. Borosilicate glass.
  • Description & Specifications
Related Products
Product Content Feedback