Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Economy Aluminum Tuning Fork
SCI_ED

Economy aluminum tuning forks are designed for sound wave lessons and exploration

$4.70 - $7.45

Specifications

Product Type Tuning Fork
Material Aluminum
For Use With (Application) Sound Wave Lessons and Exploration
View More Specs

Products 3
Catalog Number Mfr. No. Frequency Price Quantity & Availability  
Catalog Number Mfr. No. Frequency Price Quantity & Availability  
S27729
SCI_ED
View Documents
Eisco™
PH0737AA
128 Hz
Each for $4.70
Add to cart
 
S27730
SCI_ED
View Documents
Eisco™
PH0737C
320 Hz
Each for $7.45
Add to cart
 
S83614
SCI_ED
View Documents
Eisco™
PH0737H
512 Hz
Each for $7.40
Add to cart
 
Description

Description

  • Aluminum tuning fork
  • Economy priced
  • Frequency within ± 5%
Specifications

Specifications

Tuning Fork
Aluminum
Sound Wave Lessons and Exploration
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit