Learn More
Economy Micropipetters
SCI_ED
Achieve optimal pipetting. Poulten Graf™ Economy Micropipettes are used for measuring and transferring quantities of liquids from one container to another.
Specifications
Accuracy ±0.75%
View More Specs
Products 3
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Color Volume (Metric) Price Quantity & Availability  
S317181
SCI_ED
View Documents
Fischer Technical Company
E100001
Blue 100 to 1000 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S317183
SCI_ED
View Documents
Fischer Technical Company
E100005
Pink 500 to 5000 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
S317184
SCI_ED
View Documents
Fischer Technical Company
E100010
Turquoise 1000 to 10,000 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Price breakthrough in micropipetters!
Unlike most micropipetters costing hundreds of dollars, these economy micropipetters relieve strained budgets.
  • Working accuracy of ±0.75 or better
  • Normal repeatability between 0.07% and 0.34%
  • Lightweight for easy pipetting
  • Tip ejector designed for large or small, right or left hands
  • All four models are continuously variable within their range
For pipetting into or out of test tubes, choose the blue model (extended shaft).
Specifications
Specifications
±0.75%