Learn More
Economy Micropipetters
Achieve optimal pipetting. Poulten Graf™ Economy Micropipettes are used for measuring and transferring quantities of liquids from one container to another.
Manufacturer: Fischer Technical Company E100001
Description
Price breakthrough in micropipetters!
Unlike most micropipetters costing hundreds of dollars, these economy micropipetters relieve strained budgets.
• Working accuracy of ±0.75 or better
• Normal repeatability between 0.07% and 0.34%
• Lightweight for easy pipetting
• Tip ejector designed for large or small, right or left hands
• All four models are continuously variable within their range
For pipetting into or out of test tubes, choose the blue model (extended shaft).
Specifications
±0.75% | |
Blue | |
100 to 1000 μL |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.