Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Economy Polypropylene Tapes

For use indoors on smooth, flat surfaces. Brady economy polypropylene tapes are used with BMP 71 printer.

Manufacturer:  Brady™ M71C2000529

 View more versions of this product

Catalog No. 19-108-120


Add to cart

Specifications

Specifications

Safety
White
50.80 mm
BMP71 printer
50 ft.
Continuous
0.0035 in.
2 in.
Polypropylene (B-529)
1 roll
Acrylic, Permanent
Suggested ribbon series: R4400; R6000; R6900
Gloss
1
15.2 m
-40°C to +100°C
0.089 mm
5.08 cm
Economy polypropylene tape
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit