Learn More
Bel-Art™ Economy Sampling Scoops
An inexpensive and accurate measure for dry or liquid sample collection.
$18.87 - $33.70
Specifications
Bottom Shape Flat
Color White
Product Type Sampling scoop
Material FDA grade, high-impact polystyrene plastic
Sterility Non-Sterile
View More Specs
Products 4
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Price Quantity & Availability  
Catalog Number Mfr. No. Capacity Dimensions (L x W x H) Price Quantity & Availability  
14-245-55
View Documents
Bel-Art™
F369330021
2 oz., 63 mL 12.9 x 4.4 x 2.7 cm (5¹⁄₁₆ x 1¾ x 1¹⁄₁₆ in.) Pack of 12 for $18.87
Add to cart
14-245-56
View Documents
Bel-Art™
F369330022
2.7 oz., 83 mL 14.1 x 4.9 x 2.9 cm (5⁹⁄₁₆ x 1¹⁵⁄₁₆ x 1⅛ in.) Pack of 12 for $21.15
Add to cart
14-245-57
View Documents
Bel-Art™
F369330023
4 oz., 125 mL 15.9 x 5.6 x 3.2 cm (6¼ x 2³⁄₁₆ x 1¼ in.) Pack of 12 for $25.45
Add to cart
14-245-58
View Documents
Bel-Art™
F369330024
8 oz., 250 mL 18.9 x 6.8 x 4.1 cm (7⁷⁄₁₆ x 2¹¹⁄₁₆ x 1⅝ in.) Pack of 12 for $33.70
Add to cart
Description
Description
  • Capacity is listed on each white scoop
  • Leveling the sample to the flat top edge ensures a repeatable and consistent volume measure
  • Flat bottom shape and light weight allows for use as a weighing boat
  • One-piece scoops are made from durable, FDA grade, high-impact polystyrene plastic
Specifications
Specifications
Flat
Sampling scoop
Non-Sterile
White
FDA grade, high-impact polystyrene plastic