Learn More
Mystaire™ Economy Workstation Main Filter
For use with the Mystaire™ Economy Workstation (FE-2620). Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
Manufacturer:  Mystaire™ FE1021K
 View more versions of this product
Catalog No. 15-338-142
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected.
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR).
Specifications
Specifications
Main Filter
Carbon
Gas Phase Carbon
Filter
FE-2620
Formaldehyde