Learn More
Mystaire™ Economy Workstation Main Filter Available on GSA/VA Contract for Federal Government customers only.
For use with the Mystaire™ Economy Workstation (FE-2620). Mystaire™ filters are manufactured to the strictest quality standards in the industry to ensure their capability to meet your application challenges.
$295.00 - $560.00
Specifications
Material Gas Phase Carbon
Product Line Main Filter
For Use With (Equipment) FE-2620
View More Specs
Products 3
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Application) Price Quantity & Availability  
15-338-142
View Documents
Mystaire™
FE1021K
Formaldehyde
Each for $560.00
Add to cart
15-338-141 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Mystaire™
FE1021B
Hydrocarbons
Each for $295.00
Add to cart
15-338-143
View Documents
Mystaire™
FE1021J
Mineral Acids
Each for $424.00
Add to cart
Description
Description
  • Proper filtration selection is critical to ensure operator safety from potentially harmful vapors and airborne particulate.
  • Made in the U.S.A.
  • Each filter application is reviewed to ensure a proper filtration package is selected.
  • Filter performance verified by Fourier Transform Infrared Spectroscopy (FTIR).
Specifications
Specifications
Gas Phase Carbon
Main Filter
FE-2620