Learn More
EcoSpheres Products for Science Education
NASA-developed, self-contained ecosystems teeming with life.
$100.00 - $261.00
Specifications
Quantity Each
For Use With (Application) For prolonged space travel experiments, provide powerful insights about life.
Includes Active microorganisms, bright red shrimp and algae, a clear soup of filtered sea water
View More Specs
Products 4
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
S65912 Products for Science Education
View Documents
Images SI Inc
ECO01
Small Sphere Ecosphere
Each for $100.00
Add to cart
S65913 Products for Science Education
View Documents
Images SI Inc
ECO02
Small Pod Ecosphere
Each for $104.10
Add to cart
S65915 Products for Science Education
View Documents
Images SI Inc
ECO04
Large Pod Ecosphere
Each for $261.00
Add to cart
S65914 Products for Science Education
View Documents
Images SI Inc
ECO03
Medium Sphere Ecosphere
Each for $251.50
Add to cart
Description
Description
  • Completely biologically balanced
  • No cleaning, feeding or maintenance required
  • Shrimp, snails, algae and bacteria all complete their life cycles in the enclosed spheres
  • Developed for prolonged space travel experiments
Specifications
Specifications
Each
For prolonged space travel experiments, provide powerful insights about life.
Active microorganisms, bright red shrimp and algae, a clear soup of filtered sea water
Safety and Handling
Safety and Handling
ShelfLife : 2 to 7 Years