Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECSCR Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548941

Catalog No. PA548941

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Regulates endothelial chemotaxis and tube formation. Has a role in angiogenesis and apoptosis via modulation of the actin cytoskeleton and facilitation of proteasomal degradation of the apoptosis inhibitors BIRC3/IAP1 and BIRC2/IAP2.
Specifications

Specifications

ECSCR
Polyclonal
PBS with 0.09% sodium azide
Q19T08, Q3TZW0
Endothelial cell-specific chemotaxis regulator, Apoptosis regulator through modulating IAP expression, ARIA, Endothelial cell-specific molecule 2, ECSCR, ECSM2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
641700, 68545
Western Blot
Unconjugated
ECSCR
Liquid
ECSCR
KLH conjugated synthetic peptide between 137-165 amino acids from the C-terminal region of human ECSCR.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit