Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ECT2 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI2D9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270613JF549

Catalog No. N270613J549


Add to cart

Description

Description

ECT2 Monoclonal antibody specifically detects ECT2 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

ECT2
OTI2D9
Immunohistochemistry
Affinity Purified, Conjugated
ECT2
Recombinant protein fragment corresponding to amino acids 175-438 of human ECT2 (NP_060568) produced in E.coli.
Immunogen affinity purified
Phospho Specific
Primary
1894.0
Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit