Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158321
Description
ECT2 Polyclonal specifically detects ECT2 in Human samples. It is validated for Western Blot.Specifications
ECT2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q9H8V3 | |
ECT2 | |
Synthetic peptides corresponding to ECT2(epithelial cell transforming sequence 2 oncogene) The peptide sequence was selected from the middle region of ECT2. Peptide sequence PECGRQSLVELLIRPVQRLPSVALLLNDLKKHTADENPDKSTLEKAIGSL. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Guinea pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Xenopus: 92%; Equine: 92%; Bovine: 85%; Chicken: 85%; Canine: 85%; Zebrafish: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2 | |
Rabbit | |
Affinity Purified | |
RUO | |
1894 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title