Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ECT2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Specifications
Antigen | ECT2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP158321
|
Novus Biologicals
NBP158321 |
100 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
ECT2 Polyclonal specifically detects ECT2 in Human samples. It is validated for Western Blot.Specifications
ECT2 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
ARHGEF31, epithelial cell transforming sequence 2 oncogene, Epithelial cell-transforming sequence 2 oncogene, FLJ10461, MGC138291, protein ECT2 | |
ECT2 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q9H8V3 | |
1894 | |
Synthetic peptides corresponding to ECT2(epithelial cell transforming sequence 2 oncogene) The peptide sequence was selected from the middle region of ECT2. Peptide sequence PECGRQSLVELLIRPVQRLPSVALLLNDLKKHTADENPDKSTLEKAIGSL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title