Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDA/Ectodysplasin Mouse anti-Human, Alexa Fluor™ 350, Clone: 17Y41, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP221974AF350

Catalog No. NP221974A30


Add to cart

Description

Description

EDA/Ectodysplasin Monoclonal antibody specifically detects EDA/Ectodysplasin in Human samples. It is validated for ELISA.
Specifications

Specifications

EDA/Ectodysplasin
17Y41
ELISA
Conjugated, Purified
EDA
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant EDA. Mouse anti human EDA NP_001005609.
Protein G purified
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
ectodysplasin A, ED1
Mouse
IgG2
0.1 mL
Primary
1896.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit