Learn More
EDA/Ectodysplasin Mouse anti-Human, Alexa Fluor™ 594, Clone: 17Y41, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP221974AF594
Catalog No. NP221974A94
Add to cart
Description
Description
EDA/Ectodysplasin Monoclonal antibody specifically detects EDA/Ectodysplasin in Human samples. It is validated for ELISA.
Specifications
Specifications
EDA/Ectodysplasin
17Y41
ELISA
ectodysplasin A, ED1
Mouse
IgG2
Store at 4°C in the dark.
Monoclonal
Human
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
EDA
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a mouse immunized with purified recombinant EDA. Mouse anti human EDA NP_001005609.
0.1 mL
Primary
1896