Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDA/Ectodysplasin Mouse anti-Human, Mouse, Allophycocyanin/Cy7, Clone: Renzo-2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP259772APCCY7

Catalog No. N259772APC7


Add to cart

Description

Description

EDA/Ectodysplasin Monoclonal antibody specifically detects EDA/Ectodysplasin in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

EDA/Ectodysplasin
Renzo-2
Flow Cytometry
Conjugated, Purified
EDA
Recombinant human EDA-A1 (ectodysplasin-A1) extracellular domain (aa 245-391).
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Recognizes human and mouse EDA-A1 and EDA-A2.
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
ectodysplasin A, ED1
Mouse
IgG1
0.1 mL
Primary
1896.0
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit