Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDA Polyclonal Antibody
EDA Polyclonal Antibody
Supplier: Thermo Scientific PA569320
Description
EDA Polyclonal Antibody for Western Blot

Specifications
EDA | |
Polyclonal | |
Unconjugated | |
EDA | |
ECTD1; ectodermal dysplasia 1, anhidrotic; ectodermal dysplasia protein; Ectodysplasin; Ectodysplasin 1, isoform A2; ectodysplasin A; ectodysplasin-1; Ectodysplasin-A; Ectodysplasin-A, membrane form; Ectodysplasin-A, secreted form; Ed1; ED1-A1; ED1-A2; EDA; EDA 1; EDA protein; EDA protein homolog; EDA1; EDA2; Eda-A1; EDA-A2; HED; HED1; nackt; nkt; ODT1; oligodontia 1; RGD1563178; si:ch73-223d24.5; STHAGX1; Ta; tabby; Tabby protein; TNLG7C; tumor necrosis factor ligand 7C; XHED; XLHED; X-linked anhidroitic ectodermal dysplasia protein | |
Rabbit | |
Affinity Chromatography | |
RUO | |
302424 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
EDA | |
synthetic peptide directed towards the C-terminal region of rat EDA. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction