Learn More
Abnova™ EDA2R 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00060401T02
Catalog No. 89-942-742
Add to cart
Specifications
Specifications
Western Blot
293T
NM_021783
pCMV-EDA2R full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Human
EDA2R 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 32.7kDa
Transient overexpression cell lysate was tested with anti-EDA2R antibody (Mfr. No. H00060401-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)