Learn More
EDC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 177371AP
Catalog No. 50-173-1756
Add to cart
Description
Description
USP9X is an ubiquitin-specific protease involved in the removal and recycling of ubiquitin from proteins degraded in proteasomes. As a deubiquitinating enzyme, USP9x has a critical function in tight junction biogenesis by controlling the levels of the exchange factor for Arf6 (EFA6), a protein shown to facilitate tight junction formation during a narrow temporal window preceding the establishment of cell polarity. USP9X is also located on the X-chromosome but escapes X-inactivation, and defects in USP9X may cause gonadal degeneration in Turner syndrome.
Specifications
Specifications
EDC4
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3UJB9, Q3ZAV8, Q6P2E9
Autoantigen Ge 1, Autoantigen RCD 8, EDC4, enhancer of mRNA decapping 4, Ge 1, HEDLS, RCD 8
Rabbit
IgG
150 μL
-20°C
Primary
234699, 23644, 361399
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EDC4
Liquid
Edc4
EDC4 Fusion Protein Ag11784
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat