Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edelfosine, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
77286-66-9
C27H58NO6P
523.736
MHFRGQHAERHWKZ-HHHXNRCGSA-N
et-18-och3,1-o-octadecyl-2-o-methyl-sn-glycero-3-phosphocholine,1-octadecyl-2-methyl-sn-glycero-3-phosphocholine,unii-lz198hy9oe,lz198hy9oe,1-o-octadecyl-2-o-methyl-sn-glycero-3-phosphorylcholine,3,5,9-trioxa-4-phosphaheptacosan-1-aminium, 4-hydroxy-7-methoxy-n,n,n-trimethyl-, inner salt, 4-oxide, r,r-edelfosine,edelfosine, r,+-r-edelfosine
6918215
CHEBI:78653
[(2R)-2-methoxy-3-octadecoxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
CCCCCCCCCCCCCCCCCCOCC(COP(=O)([O-])OCC[N+](C)(C)C)OC
This item is not returnable. View return policy
77286-66-9
C27H58NO6P
523.736
MHFRGQHAERHWKZ-HHHXNRCGSA-N
et-18-och3,1-o-octadecyl-2-o-methyl-sn-glycero-3-phosphocholine,1-octadecyl-2-methyl-sn-glycero-3-phosphocholine,unii-lz198hy9oe,lz198hy9oe,1-o-octadecyl-2-o-methyl-sn-glycero-3-phosphorylcholine,3,5,9-trioxa-4-phosphaheptacosan-1-aminium, 4-hydroxy-7-methoxy-n,n,n-trimethyl-, inner salt, 4-oxide, r,r-edelfosine,edelfosine, r,+-r-edelfosine
6918215
CHEBI:78653
[(2R)-2-methoxy-3-octadecoxypropyl] 2-(trimethylazaniumyl)ethyl phosphate
CCCCCCCCCCCCCCCCCCOCC(COP(=O)([O-])OCC[N+](C)(C)C)OC

Selective PI-PLC inhibitor, also PAF receptor agonist

Synthetic lysophospholipid analog that selectively inhibits phosphatidylinositol phospholipase C (IC 50 = 9.6 μM in fibroblasts and adenocarcinoma cells). Also acts as an agonist at platelet-activating factor (PAF) receptors. Antitumor lipid; selectively induces apoptosis in tumor cells, sparing normal cells.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 523.73
Percent Purity >98%
Chemical Name or Material Edelfosine

Recommended Storage : Store at -20°C