Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ EDEM3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568189
Description
Predicted to react with Mouse samples.
Involved in endoplasmic reticulum-associated degradation (ERAD). Accelerates the glycoprotein ERAD by proteasomes, by catalyzing mannose trimming from Man8GlcNAc2 to Man7GlcNAc2 in the N-glycans. Seems to have alpha 1,2-mannosidase activity.
Specifications
EDEM3 | |
Polyclonal | |
Unconjugated | |
EDEM3 | |
2310050N11Rik; Alpha-1,2-mannosidase EDEM3; C1orf22; EDEM3; ER degradation enhancer, mannosidase alpha-like 3; ER degradation enhancing alpha-mannosidase like protein 3; ER degradation-enhancing alpha-mannosidase-like 3; ER degradation-enhancing alpha-mannosidase-like protein 3; ER degradation-enhancing -mannosidase-like protein 3; RGD1561496 | |
Rabbit | |
affinity chromatography | |
RUO | |
66967, 80267 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q2HXL6, Q9BZQ6 | |
EDEM3 | |
Synthesized peptide derived from human EDEM3. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction