Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetate Disodium, Dihydrate, BiotechGrade, 99-101%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6381-92-6
C10H18N2Na2O10
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
6381-92-6
C10H18N2Na2O10
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

E1058, 6381-92-6, C10H14N2Na2O8•2H2O

Spectrum™ Edetate Disodium, Dihydrate, BiotechGrade is an aminopolycarboxylic acid and a colorless, water-soluble solid. It is useful in the laboratory as a hexadentate ligand and chelating agent. The GR brand is an EMD Chemical trademark and indicates this chemical reagent is suitable for use in analytical chemistry and that the product meets or exceeds American Chemical Society (ACS) requirements.
Packaging Poly Pail
Appearance White, crystallin
Quantity 12 kg
Loss on Drying 8.7-11.4%
CAS Max % 1
Percent Purity 99 to 101%