Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetate Disodium Dihydrogen, Dihydrate, Primary Standard, 99.9-100.1%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
Packaging:
Poly Bottle
6381-92-6
C10H18N2Na2O10
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
6381-92-6
C10H18N2Na2O10
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

E1247, 6381-92-6, C10H14N2Na2O8•2H2O

Spectrum™ Edetate Disodium Dihydrogen, Dihydrate, Primary Standard is a single component organic chemical reference standard. Reference standards are particularly important for analytical chemistry since most analytical instrumentation is comparative; it requires a sample of known composition (reference standard) for accurate calibration. These reference standards are produced under stringent manufacturing procedures and differ from laboratory reagents in their certification and the traceability of the data provided.
Packaging Poly Bottle
Quantity 100 g
CAS Max % 1
Percent Purity 99.9 to 100.1%