Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edetic Acid, NF, 98-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
2.5 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
60-00-4
C10H16N2O8
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
60-00-4
C10H16N2O8
292.24
KCXVZYZYPLLWCC-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid
OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

E1208, 60-00-4, C10H16N2O8

Spectrum™ Edetic Acid, NF is used as a chelating agent in therapy and as preservative adjuncts in pharmaceutical formulations. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 98 to 100.5%
Ignition Residue 0.002
Packaging Poly Pail
Quantity 2.5 kg