Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell™ EDGE™ 48-125 Abrasion Resistant Glove

Ansell™ EDGE™ 48-125 Abrasion Resistant Glove offers excellent grip and high tactility in dry to slightly oily environments

$19.25

Specifications

Certifications/Compliance EN420:2003 + A1:2009; Category ll; REACH Compliant
Color White
For Use With (Application) Light applications
Material Polyurethane, Polyester
Product Type Glove
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-500-784 View Documents Ansell™
48125060
6 Pack of 12 for $19.25
Case of 12 PK for $183.31
Add to cart
 
19-500-785 View Documents Ansell™
48125070
7 Pack of 12 for $19.25
Case of 12 PK for $183.31
Add to cart
 
19-500-786 View Documents Ansell™
48125080
8 Pack of 12 for $19.25
Case of 12 PK for $183.31
Add to cart
 
19-500-787 View Documents Ansell™
48125090
9 Pack of 12 for $19.25
Case of 12 PK for $183.31
Add to cart
 
19-500-788 View Documents Ansell™
48125100
10 Pack of 12 for $19.25
Case of 12 PK for $183.31
Add to cart
 
19-500-789 View Documents Ansell™
48125110
11 Pack of 12 for $19.25
Case of 12 PK for $183.31
Add to cart
 
Description

Description

  • EN level 4 of abrasion resistance
  • Tear resistance
  • Good grip for improved productivity
  • Shows dirt contamination quickly
  • Seamless structure to ensure comfort and encourage safety
Specifications

Specifications

EN420:2003 + A1:2009; Category ll; REACH Compliant
Light applications
Glove
White
Polyurethane, Polyester
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit