Learn More
Ansell™ EDGE™ 48-126 Industrial Work Glove
Ansell™ EDGE™ 48-126 Industrial Work Glove combines a seamless knitted liner with elasticized knit wrist dipped into black polyurethane coating.
Manufacturer:  Ansell™ 48126070
 View more versions of this product
Catalog No. 19-500-791
Add to cart
Description
Description
  • EN level 4 abrasion resistance
  • Tear resistance
  • Good grip for improved productivity
  • High tactility in dry to slightly oily environments
  • Shows dirt less and therefore discarded less quickly
  • Seamless structure ensures comfort and encourages safety
Specifications
Specifications
Industrial Gloves
Assembly of small pieces, General handling, Packaging, Quality inspection
Polyester, Polyurethane
7
12 pairs/bag, 12 bags/carton
EN420:2003 + A1:2009; Category ll
Knit wrist
Lined
Polyester
EDGE™ 48-126
Black
REACH Compliant