Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDNRA/Endothelin R Type A, Mouse anti-Human, Clone: 485709, R&D Systems™

Mouse Monoclonal Antibody

$119.60 - $304.50

Specifications

Antigen EDNRA/Endothelin R Type A
Clone 485709
Immunogen NS0 mouse myeloma cell line transfected with human EDNRA/Endothelin R Type A, Accession # NP_001948
Host Species Mouse
Primary or Secondary Primary
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB65381100
View Documents
R&D Systems
MAB65381100
100 ug
Each for $304.50
Add to cart
 
MAB65381SP
View Documents
R&D Systems
MAB65381SP
25 ug
Each for $119.60
Add to cart
 
Description

Description

EDNRA/Endothelin R Type A Monoclonal antibody specifically detects EDNRA/Endothelin R Type A in Human Samples. It is validated for Flow Cytometry, CyTOF.
Specifications

Specifications

EDNRA/Endothelin R Type A
NS0 mouse myeloma cell line transfected with human EDNRA/Endothelin R Type A, Accession # NP_001948
Primary
Human
Unconjugated
1909
Purified
Flow Cytometry, CyTOF
RUO
485709
Mouse
Monoclonal
IgG2b
EDNRA, endothelin receptor type A, endothelin-1 receptor, endothelin-1-specific receptor, ET1-specific type, ETAR, ETRA, G protein-coupled receptor, hET-AR
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit