Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human EDNRB/Endothelin R Type B Alexa Fluor 405-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Supplier: R&D Systems FAB4496V100UG
This item is not returnable.
View return policy
Description
EDNRB/Endothelin R Type B Monoclonal specifically detects EDNRB/Endothelin R Type B in Human samples. It is validated for Flow Cytometry.Specifications
EDNRB/Endothelin R Type B | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
ABCDS, EDNRB, endothelin B receptor, Endothelin receptor non-selective type, endothelin receptor type B, ETB, ET-B, ETBR, ET-BR, ETRB, HSCR, HSCR2, WS4A | |
NS0 mouse myeloma cell line transfected with human EDNRB/Endothelin R Type B, Accession # P24530 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
671917 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P24530 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1910 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction