Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDNRB/Endothelin R Type B Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
$175.82 - $436.00
Specifications
Antigen | EDNRB/Endothelin R Type B |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP130599SS
|
Novus Biologicals
NBP130599SS |
0.1 mg |
Each for $175.82
|
|
NBP130599
|
Novus Biologicals
NBP130599 |
0.5 mg |
Each for $436.00
|
|
Description
EDNRB/Endothelin R Type B Polyclonal specifically detects EDNRB/Endothelin R Type B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
EDNRB/Endothelin R Type B | |
Western Blot 10-50 ug/ml, Immunohistochemistry 10-50 ug/ml, Immunohistochemistry-Paraffin 10-50 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
P48302, P21451 | |
1910 | |
A synthetic peptide from the c-terminal of human Endothelin B receptor (EDNRB) conjugated to an immunogenic carrier protein was used as the immunogen. The peptide is homologous in rat and mouse. | |
Primary | |
Reconstitute with 0.5 ml sterilized water. | |
Appears to be specific for EDNRB. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
GPCR, Neuroscience, Signal Transduction | |
Sterilized water with No Preservative | |
ABCDS, endothelin B receptor, Endothelin receptor non-selective type, endothelin receptor type B, ETB, ET-B, ETBR, ET-BR, ETRB, HSCR, HSCR2, WS4A | |
EDNRB | |
IgG | |
IgG purified | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title