Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
EDNRB/EndothelinRTypeB Rabbit anti-Human, Clone: JM74-10, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266817
Description
EDNRB/EndothelinRTypeB Monoclonal antibody specifically detects EDNRB/EndothelinRTypeB in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
EDNRB/EndothelinRTypeB | |
JM74-10 | |
Unconjugated | |
Purified | |
ABCDS, endothelin B receptor, Endothelin receptor non-selective type, endothelin receptor type B, ETB, ET-B, ETBR, ET-BR, ETRB, HSCR, HSCR2, WS4A | |
EDNRB | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
GPCR, Neuroscience, Signal Transduction | |
Human,Zebrafish |
Immunoprecipitation,Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:10 - 1:50 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
1910 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title