Learn More
Enzo Life Sciences Endothelin receptor ETB polyclonal antibody (100µg)
Supplier: Enzo Life Sciences ADI905791100
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat endothelin receptor ETB. Species reactivity: Rat. Applications: IHC, WB. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.01% sodium azide.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.