Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDR1/PHC1 Mouse anti-Human, FITC, Clone: 1F3F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237371F

Catalog No. NBP237371F

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

EDR1/PHC1
1F3F3
Flow Cytometry, ELISA
Purified
PHC1
Purified recombinant fragment of human PHC1 (AA: 758-1004) expressed in E. Coli.
Protein A or G purified
Core ESC Like Genes, Stem Cell Markers
Primary
1911
ELISA, Flow Cytometry
FITC
PBS with 0.05% Sodium Azide
early development regulator 1 (homolog of polyhomeotic 1), Early development regulatory protein 1, EDR1, hPH1, PH1, polyhomeotic homolog 1 (Drosophila), polyhomeotic-like 1, polyhomeotic-like 1 (Drosophila), polyhomeotic-like protein 1, RAE28
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit