Learn More
EDR1/PHC1 Mouse anti-Human, HRP, Clone: 1F3F3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237371H
Catalog No. NBP237371H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
EDR1/PHC1
1F3F3
Western Blot, ELISA
Purified
PHC1
Purified recombinant fragment of human PHC1 (AA: 758-1004) expressed in E. Coli.
Protein A or G purified
Core ESC Like Genes, Stem Cell Markers
Primary
1911
ELISA, Western Blot
HRP
PBS with No Preservative
early development regulator 1 (homolog of polyhomeotic 1), Early development regulatory protein 1, EDR1, hPH1, PH1, polyhomeotic homolog 1 (Drosophila), polyhomeotic-like 1, polyhomeotic-like 1 (Drosophila), polyhomeotic-like protein 1, RAE28
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human