Learn More
EDRF1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 218831AP
Catalog No. 50-173-3623
Add to cart
Description
Description
This gene may play a role in erythroid cell differentiation. The encoded protein inhibits DNA binding of the erythroid transcription factor GATA-1 and may regulate the expression of alpha-globin and gamma-globin. Alternative splicing results in multiple transcript variants.
Specifications
Specifications
EDRF1
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3B7T1, Q6GQV7
C10orf137, EDRF1, RP11 383C5.6
Rabbit
IgG
150 μL
-20°C
Primary
214764, 26098, 309069
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
EDRF1
Liquid
EDRF1
EDRF1 Fusion Protein Ag16441
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat