Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Buffer, No Magnesium Water Hardness Buffer, Reagents

Catalog No. 01335335
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
Catalog No. Quantity
01-335-335 500 mL
01-335-338 10 L
01-335-337 1 L
01-335-336 4 L
4 options

Catalog No. 01-335-335

Supplier: Reagents Holdings Llc CE252000500A

Only null left
Add to Cart
Add to Cart

EDTA Buffer, No Magnesium Water Hardness Buffer, Reagent, use for Metals analysis (EDTA method). Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Buffer
CAS 7732-18-5
Color Colorless
For Use With (Application) Metals Analysis (EDTA method)
pH 10
Physical Form Liquid
Quantity 500 mL
Packaging Poly Bottle
Sterility Non-sterile
Synonym Water Hardness Buffer
Solubility Information Completely miscible
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.