Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Disodium, 0.01 M, For Water Hardness, APHA, Solution, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
6381-92-6
6381-92-6
Concentration 0.01 M ± 0.0002 M
Grade Chemical Solution
Packaging Poly Bottle
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.