Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Disodium, 10% (w/v) Solution, Spectrum™

Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Poly Bottle
6381-92-6,7732-18-5
C10H18N2Na2O10
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O
6381-92-6,7732-18-5
C10H18N2Na2O10
372.24
OVBJJZOQPCKUOR-UHFFFAOYSA-L
disodium 2-({2-[(carboxylatomethyl)(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetate dihydrate
O.O.[Na+].[Na+].OC(=O)CN(CCN(CC(O)=O)CC([O-])=O)CC([O-])=O

E-091, 6381-92-6

Spectrum™ EDTA Disodium, 10 Percent (w/v) Solution , is generally used as a gentle non-enzymatic cell dissociation reagent, a chelating reagent and in biological and electrophoresis buffer systems.
Packaging Poly Bottle
Quantity 125 mL
CAS Max % 0.1,0.9
Percent Purity 10% +/ to 0.5