Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Disodium Magnesium Salt, Tetrahydrate, J.T.Baker™

Click to view available options
Quantity:
100g
5kg
Packaging:
Poly Bottle
Poly Pail
29932-54-5
C10H20MgN2Na2O12
430.557
NTLBRFLUZXNZAU-UHFFFAOYSA-J
ethylenediaminetetraacetic acid disodium magnesium salt tetrahydrate
131700215
magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Mg+2]
29932-54-5
C10H20MgN2Na2O12
430.557
NTLBRFLUZXNZAU-UHFFFAOYSA-J
ethylenediaminetetraacetic acid disodium magnesium salt tetrahydrate
131700215
magnesium;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Mg+2]
Quantity 100g
Packaging Poly Bottle
Formula Weight 430.56
Chemical Name or Material EDTA Disodium Magnesium Salt, Tetrahydrate