Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDTA Disodium Salt 5% (w/v) Solution, For Nitrogen (Ammonia)For Nitrogen (Organic)APHA, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
Packaging:
Poly Bottle
6381-92-6
6381-92-6
Grade Chemical Solution
Packaging Poly Bottle
Quantity 1 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.