Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ EDTA Disodium Salt Dihydrate, 99-101.0%, MB Grade, Ultrapure, Thermo Scientific™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Plastic bottle
6381-92-6
C10H18N2Na2O10
372.238
MFCD00150037
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]
6381-92-6
C10H18N2Na2O10
372.238
MFCD00150037
OVBJJZOQPCKUOR-UHFFFAOYSA-L
edta disodium salt,cal-ex decalcifier,buffer solution, ph 10.00,sodium di ethylenediamine tetraacetate dihydrate,ethylenediamine tetraacetic acid, disodium salt dihydrate,ethylenediamine tetraacetic acid, disodium salt, standard solution,sodium di ethylenediamine tetraacetate standard solution,ethylenedinitrilo tetraacetic acid disodium, dihydrate, reagent, acs
44120005
disodium;2-[2-[bis(carboxymethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;dihydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)O)CC(=O)O.O.O.[Na+].[Na+]

Chelating agent used in many enzyme buffers and as enzyme inactivator at high concentrations

Formula Weight 372.24
Physical Form Crystalline Powder
Percent Purity 99.0 to 101.0%
Grade Molecular Biology
Packaging Plastic bottle
Melting Point 248°C
Quantity 500g
Chemical Name or Material Ethylenediaminetetraacetic Acid Disodium Salt Dihydrate